Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018815.2 | NP_061285.2 | 4 | 539 | ||
GenBank mRNA | AF113751.1 | - | 4 | 507 | ||
AK053417.1 | - | 4 | 540 | |||
AK157272.1 | - | 4 | 531 | |||
AK173056.1 | - | 4 | 493 | |||
AK202156.1 | - | 231 | ||||
BC052468.1 | - | 4 | 532 | 6835038 |