Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286260.1 | NP_001273189.1 | 3 | 777 | ||
NM_018823.2 | NP_061293.2 | 4 | 831 | |||
NM_133957.3 | NP_598718.2 | 5 | 893 | |||
GenBank mRNA | AF162853.1 | - | 3 | 688 | ||
AF200687.1 | - | 4 | 466 | |||
AF228705.1 | - | 1 | 531 | |||
AF228706.1 | - | 1 | 531 | |||
AF369980.1 | - | 5 | 532 | |||
AF453571.2 | - | 5 | 501 | |||
AK134022.1 | - | 5 | 897 | |||
AK136696.1 | - | 1 | 2326 |