Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018830.1 | NP_061300.1 | 6 | 713 | ||
GenBank mRNA | AB037111.1 | - | 7 | 998 | ||
AB037181.1 | - | 6 | 713 | |||
AK034493.1 | - | 6 | 944 | |||
AK041937.1 | - | 6 | 902 | |||
AK046540.1 | - | 6 | 907 | |||
AK047692.1 | - | 6 | 901 | |||
AK080951.1 | - | 6 | 890 | |||
AK166100.1 | - | 6 | 961 | |||
BC137898.1 | - | 6 | 696 | 8860917 | ||
BC145016.1 | - | 6 | 699 | 9053564 |