Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207678.1 | NP_997561.1 | 11 | 1537 | ||
GenBank mRNA | AB041605.1 | - | 11 | 1514 | ||
AK008585.1 | - | 6 | 531 | |||
AK048244.1 | - | 11 | 3317 | |||
AK193127.1 | - | 1 | 22 | |||
AY337018.1 | - | 11 | 1520 | |||
BC003773.1 | - | 4 | 660 | |||
BC023747.1 | - | 11 | 3305 | |||
BC083055.1 | - | 4 | 524 | |||
BC132295.1 | - | 11 | 1529 | 40130572 | ||
U37351.1 | - | 4 | 543 |