Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agpat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163379.1 | NP_001156851.1 | 4 | 746 | ||
NM_018862.3 | NP_061350.3 | 4 | 498 | |||
GenBank mRNA | AB005623.1 | - | 4 | 744 | ||
AK086281.1 | - | 4 | 688 | |||
BC009651.1 | - | 4 | 498 | 3487719 |