Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018768.2 | NP_061238.1 | 3 | 182 | ||
GenBank mRNA | AB040054.1 | - | 3 | 179 | ||
AF036716.1 | - | 3 | 169 | |||
AF093064.1 | - | 3 | 191 | |||
AK003297.1 | - | 3 | 208 | |||
AK011376.1 | - | 3 | 196 | |||
AK016271.1 | - | 3 | 208 | |||
AK035105.1 | - | 3 | 186 | |||
BC038186.1 | - | 3 | 181 | |||
BC048167.1 | - | 3 | 182 | 6821351 | ||
BC048479.1 | - | 3 | 207 | 6774827 | ||
BC061118.1 | - | 3 | 207 | 30295878 |