Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dfna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018769.3 | NP_061239.1 | 10 | 1621 | ||
GenBank mRNA | AF073309.1 | - | 10 | 1622 | ||
AK016561.1 | - | 1989 | ||||
AK171278.1 | - | 10 | 1620 | |||
BC051962.1 | - | 10 | 1680 | |||
BC086618.1 | - | 10 | 1680 | |||
BC132303.1 | - | 9 | 1592 | 40130580 |