Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018771.3 | NP_061241.1 | 1611 | |||
GenBank mRNA | AF061263.1 | - | 1547 | |||
AF104358.1 | - | 1590 | ||||
AK202779.1 | - | 300 | ||||
BC003490.1 | - | 1611 | 2811749 |