Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018778.3 | NP_061248.1 | 1 | 1662 | ||
GenBank mRNA | AK133810.1 | - | 1 | 1662 | ||
BC003868.1 | - | 1 | 1613 | 3490218 |