Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018782.2 | NP_061252.2 | 16 | 2743 | ||
GenBank mRNA | AB015595.1 | - | 16 | 2745 | ||
AF146525.1 | - | 14 | 2430 |
Set Membership: |
G-protein coupled receptor |