Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018784.2 | NP_061254.1 | 7 | 878 | ||
GenBank mRNA | AB063326.1 | - | 5 | 381 | ||
AF119390.1 | - | 7 | 878 | |||
AK028442.1 | - | 7 | 697 | |||
AK033562.1 | - | 7 | 765 | |||
AK041173.1 | - | 7 | 921 | |||
AK048950.1 | - | 7 | 922 | |||
AK162619.1 | - | 8 | 858 | |||
AK197012.1 | - | 3 | 154 | |||
AK207906.1 | - | 3 | 154 | |||
BC052338.1 | - | 6 | 639 | 30061291 |