Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039574.2 | NP_001034663.1 | 2 | 2384 | ||
NM_004978.4 | NP_004969.2 | 2 | 2384 | |||
NR_036437.1 | 2 | 2384 | ||||
XM_006710625.3 | XP_006710688.1 | 2 | 2228 | |||
XM_006710627.3 | XP_006710690.1 | 2 | 2228 | |||
XM_011541401.2 | XP_011539703.1 | 2 | 2228 | |||
GenBank mRNA | AK310193.1 | - | 2 | 1809 | ||
BC101769.1 | - | 2 | 1655 | 8069275 | ||
BX098184.1 | - | 1 | 46 | |||
BX537638.1 | - | 1 | 4172 | |||
CK823243.1 | - | 1 | 452 | |||
DA936036.1 | - | 1 | 337 | |||
M64676.1 | - | 2 | 1754 |