Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205312.1 | NP_001192241.1 | 25 | 4675 | ||
NM_018793.2 | NP_061263.2 | 25 | 4606 | |||
GenBank mRNA | AF173032.1 | - | 25 | 4541 | ||
AK154438.1 | - | 25 | 4633 | |||
AK155923.1 | - | 25 | 4518 | |||
AK159303.1 | - | 25 | 4575 | |||
AK167726.1 | - | 25 | 4585 | |||
AK171609.1 | - | 25 | 4518 | |||
AK217061.1 | - | 1 | 72 | |||
BC019789.1 | - | 9 | 2019 | |||
BC053021.1 | - | 19 | 4246 | |||
BC094240.1 | - | 24 | 4502 | 30610201 |