Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285857.1 | NP_001272786.1 | 9 | 1233 | ||
NM_001285858.1 | NP_001272787.1 | 9 | 1244 | |||
NM_001285861.1 | NP_001272790.1 | 9 | 1234 | |||
NM_018802.5 | NP_061272.2 | 9 | 1355 | |||
GenBank mRNA | AB026805.1 | - | 9 | 1234 | ||
AK075832.1 | - | 9 | 1356 | |||
BC100747.1 | - | 6 | 1068 | 40045988 | ||
BC100748.1 | - | 9 | 1209 | 40045989 | ||
BC103602.1 | - | 9 | 1209 | 40045982 | ||
BC103665.1 | - | 9 | 1220 | 40045986 | ||
U20107.1 | - | 7 | 1015 |