Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018814.3 | NP_061284.2 | 27, 28 | 5465 | ||
GenBank mRNA | AF096286.1 | - | 21, 22 | 3295 | ||
AK014387.1 | - | 1, 2 | 153 | |||
AK129221.1 | - | 22, 23 | 3345 | |||
AK154122.1 | - | 8, 9 | 1193 | |||
AK170320.1 | - | 8, 9 | 1193 | |||
BC057333.1 | - | 20, 21 | 3208 | |||
BC066849.1 | - | 22, 23 | 3130 | |||
BC098370.1 | - | 22, 23 | 3055 |