Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF234838.1 | - | 3, 4 | 1101 | ||
AK017604.1 | - | 3, 4 | 1071 | |||
AK140319.1 | - | 3, 4 | 1067 | |||
AK140325.1 | - | 3, 4 | 1067 | |||
BC049777.1 | - | 3, 4 | 1052 | |||
BC057334.1 | - | 3, 4 | 1119 | 6811850 |