Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018831.4 | NP_061301.3 | 3 | 1641 | ||
GenBank mRNA | AF241240.1 | - | 2 | 1112 | ||
AK220214.1 | - | 2 | 869 | |||
BC031858.1 | - | 1 | 532 | |||
BC072667.1 | - | 2 | 1057 | 30629417 | ||
BC076617.1 | - | 3 | 1641 |