Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019388.3 | NP_062261.3 | 3 | 256 | ||
GenBank mRNA | AF065897.1 | - | 3 | 172 | ||
AF065898.1 | - | 3 | 172 | |||
AF065899.1 | - | 3 | 172 | |||
AF065900.1 | - | 3 | 172 | |||
AK079513.1 | - | 3 | 257 | |||
AK170054.1 | - | 3 | 257 | |||
AK172548.1 | - | 3 | 259 | |||
BC013807.1 | - | 3 | 230 | 4008635 | ||
L25606.1 | - | 3 | 238 | |||
S70108.1 | - | 3 | 221 |