Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019394.3 | NP_062267.2 | 1, 2 | 226 | ||
GenBank mRNA | AK135151.1 | - | 1, 2 | 205 | ||
BC009815.1 | - | 1, 2 | 195 | |||
BE864464.1 | - | 1, 2 | 226 | |||
CK794645.1 | - | 1, 2 | 220 | |||
X94322.1 | - | 1, 2 | 231 |