Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019412.2 | NP_062285.1 | 5 | 414 | ||
NM_198048.2 | NP_932165.2 | 5 | 414 | |||
GenBank mRNA | AJ222968.1 | - | 5 | 413 | ||
AJ222969.1 | - | 5 | 413 | |||
AK132429.1 | - | 1 | 257 | |||
AK173220.1 | - | 1 | 211 | |||
BC068135.1 | - | 1 | 227 | 30354483 | ||
CJ045218.1 | - | 5 | 417 |