Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019435.4 | NP_062308.2 | 3 | 783 | ||
GenBank mRNA | AK028121.1 | - | 3 | 504 | ||
BC027265.1 | - | 3 | 776 | 3589704 | ||
BC096047.1 | - | 3 | 479 | |||
Y08702.1 | - | 3 | 476 |