Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019439.3 | NP_062312.3 | 2, 3 | 414 | ||
GenBank mRNA | AF008649.1 | - | 2, 3 | 298 | ||
AF114168.2 | - | 2, 3 | 312 | |||
AK134754.1 | - | 1, 2 | 208 | |||
AK156397.1 | - | 2, 3 | 361 | |||
AK159584.1 | - | 2, 3 | 353 | |||
BC054735.1 | - | 2, 3 | 414 | 6401243 |
Set Membership: |
G-protein coupled receptor |