Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018866.2 | NP_061354.1 | 4 | 635 | ||
GenBank mRNA | AF044196.1 | - | 4 | 635 | ||
AK156025.1 | - | 4 | 649 | |||
AK160241.1 | - | 4 | 651 | |||
BC012965.1 | - | 4 | 634 | 4217539 |