Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018870.3 | NP_061358.1 | 2 | 623 | ||
GenBank mRNA | AF029843.1 | - | 2 | 560 | ||
AK146703.1 | - | 2 | 550 | |||
AK169019.1 | - | 2 | 550 | |||
BC010750.1 | - | 2 | 528 | 4235174 |