Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfkb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177369.1 | NP_001170840.1 | 6, 7 | 547 | ||
NM_001177370.1 | NP_001170841.1 | 5, 6 | 463 | |||
NM_019408.3 | NP_062281.1 | 5, 6 | 698 | |||
GenBank mRNA | AF155372.1 | - | 6, 7 | 625 | ||
AF155373.1 | - | 6, 7 | 662 | |||
AK134081.1 | - | 6, 7 | 966 | |||
AK137680.1 | - | 6, 7 | 537 | |||
AK144994.1 | - | 6, 7 | 540 | |||
AK145312.1 | - | 5, 6 | 664 | |||
AK148160.1 | - | 6, 7 | 535 | |||
AK150012.1 | - | 6, 7 | 526 | |||
AK153412.1 | - | 1, 2 | 107 | |||
AK193270.1 | - | 1, 2 | 127 | |||
AK216302.1 | - | 1, 2 | 127 | |||
BC027423.1 | - | 5, 6 | 464 | 3672913 | ||
DQ020182.1 | - | 5, 6 | 395 | |||
DQ020183.1 | - | 4, 5 | 516 | |||
DQ020184.1 | - | 4, 5 | 516 | |||
DQ020185.1 | - | 5, 6 | 395 |