Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf7ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019426.2 | NP_062299.2 | 2 | 1286 | ||
GenBank mRNA | AJ132702.1 | - | 2 | 1279 | ||
AK134252.1 | - | 2 | 1293 | |||
AK167542.1 | - | 3 | 1332 | |||
BC022714.1 | - | 2 | 1279 | |||
BC037617.1 | - | 1288 | ||||
BC048827.1 | - | 3 | 1447 | |||
BC052460.1 | - | 2 | 1286 | 6834550 | ||
BC060246.1 | - | 2 | 1269 | 5705945 |