Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm3ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019434.2 | NP_062307.2 | 2 | 752 | ||
GenBank mRNA | AJ006590.1 | - | 2 | 785 | ||
AK051901.1 | - | 2 | 783 | |||
AK122313.1 | - | 2 | 796 | |||
BC052452.1 | - | 2 | 752 | 6409827 |