Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019437.3 | NP_062310.1 | 4 | 721 | ||
GenBank mRNA | AK002806.1 | - | 4 | 682 | ||
AK008352.1 | - | 4 | 651 | |||
AK010607.1 | - | 4 | 650 | |||
BC033521.1 | - | 4 | 623 | 4527980 | ||
BC051021.1 | - | 4 | 632 | 6315425 |