Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019439.3 | NP_062312.3 | 3 | 479 | ||
GenBank mRNA | AF008649.1 | - | 3 | 363 | ||
AF114168.2 | - | 3 | 377 | |||
AK134754.1 | - | 2 | 273 | |||
AK156397.1 | - | 3 | 426 | |||
AK159584.1 | - | 3 | 418 | |||
BC054735.1 | - | 3 | 479 | 6401243 |
Set Membership: |
G-protein coupled receptor |