Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081695.2 | NP_001075164.1 | 4 | 429 | ||
NM_001284197.1 | NP_001271126.1 | 2 | 270 | |||
NM_019448.4 | NP_062321.1 | 3 | 283 | |||
GenBank mRNA | AF220524.1 | - | 2 | 214 | ||
AJ404467.1 | - | 2 | 255 | |||
AK010434.1 | - | 3 | 284 | |||
AV473569.1 | - | 2 | 270 | |||
BC083147.1 | - | 2 | 268 | 30447906 | ||
EF051624.1 | - | 4 | 429 |