Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281954.1 | NP_001268883.1 | 3 | 324 | ||
NM_019465.4 | NP_062338.3 | 3 | 324 | |||
GenBank mRNA | AF001104.1 | - | 3 | 282 | ||
AK005719.1 | - | 1 | 47 | |||
BC117715.1 | - | 3 | 282 | |||
BC117716.1 | - | 3 | 282 | 40088099 |