Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081549.2 | NP_001075018.1 | 2 | 381 | ||
NM_019466.4 | NP_062339.2 | 2 | 227 | |||
GenBank mRNA | AF237789.1 | - | 2 | 131 | ||
AF237790.1 | - | 2 | 131 | |||
AF260717.1 | - | 2 | 145 | |||
AF263239.1 | - | 2 | 161 | |||
AF263240.1 | - | 2 | 244 | |||
AF282255.1 | - | 2 | 138 | |||
AK010696.1 | - | 98 | ||||
AK077283.1 | - | 2 | 227 | |||
AK146764.1 | - | 2 | 356 | |||
AY325904.1 | - | 2 | 337 | |||
BC013551.1 | - | 2 | 207 | 4236038 | ||
BY276032.1 | - | 2 | 382 |