Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018868.2 | NP_061356.2 | 6, 7 | 676 | ||
GenBank mRNA | AF053232.1 | - | 6, 7 | 581 | ||
AK044216.1 | - | 673 | ||||
AK133959.1 | - | 6, 7 | 674 | |||
AK134286.1 | - | 6, 7 | 677 | |||
BC076604.1 | - | 6, 7 | 677 | 30630088 | ||
BC085135.1 | - | 6, 7 | 646 | 5718488 |