Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198894.1 | NP_001185823.1 | 6, 7 | 1228 | ||
NM_018882.3 | NP_061370.2 | 6, 7 | 1178 | |||
GenBank mRNA | AF166382.1 | - | 6, 7 | 1228 | ||
AK087268.1 | - | 6, 7 | 980 | |||
AK133678.1 | - | 6, 7 | 945 | |||
AK141886.1 | - | 6, 7 | 1207 | |||
AK141894.1 | - | 6, 7 | 991 | |||
AK164776.1 | - | 6, 7 | 1003 | |||
AK167547.1 | - | 5, 6 | 932 | |||
AK182857.1 | - | 1, 2 | 65 | |||
BC034678.1 | - | 6, 7 | 974 | 3709247 |
Set Membership: |
G-protein coupled receptor |