Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006347.3 | NP_006338.1 | 5 | 288 | ||
XM_005270362.1 | XP_005270419.1 | 5 | 242 | |||
XM_005270366.3 | XP_005270423.1 | 5 | 895 | |||
XM_011540508.1 | XP_011538810.1 | 5 | 360 | |||
XM_017000057.1 | XP_016855546.1 | 5 | 242 | |||
XM_017000058.1 | XP_016855547.1 | 5 | 1014 | |||
XM_017000059.1 | XP_016855548.1 | 5 | 360 | |||
GenBank mRNA | AF016371.1 | - | 5 | 255 | ||
AF036331.1 | - | 5 | 231 | |||
AK294288.1 | - | 3 | 517 | |||
AK309438.1 | - | 5 | 356 | |||
AK310052.1 | - | 4 | 511 | |||
AK311979.1 | - | 5 | 241 | |||
BC003412.1 | - | 5 | 240 | 3451034 | ||
BT019343.1 | - | 5 | 218 | |||
BU955702.1 | - | 5 | 297 | |||
CR541899.1 | - | 5 | 218 | |||
CR541920.1 | - | 5 | 218 |