Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042537.1 | NP_001036002.1 | 5 | 798 | ||
NM_001177651.1 | NP_001171122.1 | 6 | 676 | |||
NM_006359.2 | NP_006350.1 | 5 | 702 | |||
XM_006724726.3 | XP_006724789.1 | 6 | 792 | |||
XM_011531243.2 | XP_011529545.1 | 6 | 556 | |||
XM_017029223.1 | XP_016884712.1 | 6 | 809 | |||
XM_017029224.1 | XP_016884713.1 | 6 | 939 | |||
XM_017029225.1 | XP_016884714.1 | 6 | 576 | |||
GenBank mRNA | AB074255.1 | - | 5 | 722 | ||
AF030409.1 | - | 5 | 661 | |||
AK289775.1 | - | 5 | 722 | |||
AK296444.1 | - | 5 | 461 | |||
AK297218.1 | - | 4 | 248 | |||
AK300475.1 | - | 6 | 667 | |||
AK314562.1 | - | 5 | 790 | |||
AK316255.1 | - | 6 | 667 | |||
BC035029.1 | - | 5 | 760 | 5264865 | ||
BC049169.1 | - | 5 | 722 | 5312949 | ||
D87743.1 | - | 5 | 617 |