Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019420.2 | NP_062293.1 | 1 | 1561 | ||
GenBank mRNA | AF082504.1 | - | 1 | 1554 | ||
AK007600.1 | - | 1 | 1549 | |||
AK166333.1 | - | 1 | 1513 |