Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Barhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164186.1 | NP_001157658.1 | 4 | 3039 | ||
NM_019446.4 | NP_062319.1 | 3 | 3322 | |||
GenBank mRNA | AB043980.1 | - | 3 | 3337 | ||
AK036312.1 | - | 3 | 2775 | |||
AK048063.1 | - | 3 | 2175 |