Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019455.4 | NP_062328.3 | 3 | 247 | ||
GenBank mRNA | AK020246.1 | - | 3 | 248 | ||
AK039392.1 | - | 3 | 248 | |||
BC116735.1 | - | 3 | 215 | 40126054 | ||
BC116737.1 | - | 3 | 215 | 40126056 | ||
D82072.1 | - | 3 | 425 |