Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019458.3 | NP_062331.2 | 11 | 1144 | ||
GenBank mRNA | AB041615.1 | - | 11 | 1162 | ||
AK017762.1 | - | 11 | 1214 | |||
AK134857.1 | - | 11 | 1157 | |||
AK148538.1 | - | 11 | 1179 | |||
AK151665.1 | - | 11 | 1190 | |||
AK152778.1 | - | 11 | 1160 | |||
AK152836.1 | - | 11 | 1190 | |||
BC010317.1 | - | 10 | 1188 | |||
BC029843.1 | - | 11 | 1144 | 2651081 | ||
BC083337.1 | - | 11 | 1125 | 5701517 |