Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019488.4 | NP_062361.1 | 10 | 1441 | ||
GenBank mRNA | AF232061.2 | - | 10 | 1398 | ||
AJ245936.1 | - | 10 | 1418 | |||
AK081806.1 | - | 10 | 1458 | |||
AK141193.1 | - | 6 | 2481 | |||
AK155792.1 | - | 10 | 1471 | |||
AK170319.1 | - | 10 | 1454 | |||
AY856043.1 | - | 10 | 1402 | |||
AY856044.1 | - | 9 | 1293 | |||
AY856045.1 | - | 8 | 1122 | |||
AY856046.1 | - | 6 | 823 | |||
BC021758.1 | - | 6 | 767 | |||
BC090993.1 | - | 10 | 1441 | 6751763 | ||
Y17802.1 | - | 10 | 1423 |