Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019510.2 | NP_062383.2 | 2 | 1017 | ||
GenBank mRNA | AF190645.1 | - | 1 | 607 | ||
AK032317.1 | - | 2 | 1018 | |||
AK080619.1 | - | 2 | 1005 | |||
AK082063.1 | - | 2 | 980 | |||
BC108988.2 | - | 1 | 606 | |||
BC108989.2 | - | 1 | 607 | 40047766 | ||
FJ207475.1 | - | 1 | 607 | |||
FJ207476.1 | - | 1 | 607 |