Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019547.2 | NP_062420.2 | 1 | 227 | ||
GenBank mRNA | AI573584.1 | - | 1 | 245 | ||
AK146026.1 | - | 1 | 201 | |||
AK162141.1 | - | 1 | 200 | |||
AK166609.1 | - | 1 | 228 | |||
AK172625.1 | - | 1 | 198 | |||
BC006687.1 | - | 1 | 216 | |||
BC085307.1 | - | 1 | 171 | 6396103 |