Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012128.3 | NP_036260.2 | 2 | 307 | ||
NR_024602.1 | 2 | 307 | ||||
XM_011541015.2 | XP_011539317.1 | 2 | 126 | |||
GenBank mRNA | AF127035.1 | - | 2 | 291 | ||
AK000072.1 | - | 2 | 305 | |||
AK293766.1 | - | 2 | 140 | |||
AK307785.1 | - | 2 | 251 | |||
AK307898.1 | - | 2 | 169 | |||
AK314430.1 | - | 2 | 303 | |||
AY358470.1 | - | 2 | 287 | |||
BC113687.1 | - | 2 | 302 | 8322739 | ||
BC113689.1 | - | 2 | 301 | 8322741 | ||
DA668017.1 | - | 2 | 308 | |||
DA761219.1 | - | 2 | 369 |