Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019581.3 | NP_062527.2 | 1 | 257 | ||
GenBank mRNA | AF168991.1 | - | 1 | 162 | ||
AK008731.1 | - | 1 | 129 | |||
AK146415.1 | - | 1 | 207 | |||
AK148020.1 | - | 1 | 178 | |||
BC049089.1 | - | 1 | 218 | 5720710 | ||
BC051958.1 | - | 1 | 218 | 5720710 |