Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019487.3 | NP_062360.1 | 2, 3 | 512 | ||
GenBank mRNA | AF117614.1 | - | 2, 3 | 442 | ||
AK038452.1 | - | 2, 3 | 501 | |||
AK078041.1 | - | 492 | ||||
AK078130.1 | - | 2, 3 | 512 | |||
BC006898.1 | - | 2, 3 | 323 | 3599858 |