Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019503.4 | NP_062376.2 | 5 | 294 | ||
NM_052991.4 | NP_443717.1 | 5 | 277 | |||
NM_052992.3 | NP_443718.1 | 5 | 358 | |||
NM_194321.2 | NP_919302.1 | 5 | 341 | |||
GenBank mRNA | AA462069.1 | - | 5 | 226 | ||
AA466595.1 | - | 5 | 294 | |||
AF089734.1 | - | 5 | 220 | |||
AK002585.1 | - | 5 | 281 | |||
AK003504.1 | - | 1 | 76 | |||
BC024671.1 | - | 5 | 231 | 4208962 | ||
BY064152.1 | - | 5 | 293 | |||
BY132051.1 | - | 5 | 263 | |||
CA459152.1 | - | 5 | 299 | |||
CK793316.1 | - | 5 | 276 | |||
W16239.1 | - | 5 | 257 | |||
W98524.1 | - | 5 | 272 |