Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabggta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019519.2 | NP_062392.1 | 5 | 824 | ||
GenBank mRNA | AF127656.1 | - | 6 | 672 | ||
AF127657.1 | - | 5 | 615 | |||
AF127658.1 | - | 5 | 824 | |||
AF127659.1 | - | 5 | 824 | |||
AF127660.1 | - | 6 | 672 | |||
AF127661.1 | - | 5 | 615 | |||
AF127662.1 | - | 6 | 743 | |||
AK002625.1 | - | 6 | 606 | |||
AK146917.1 | - | 6 | 695 | |||
BC012214.1 | - | 6 | 677 | 3967218 |