Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhog | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019566.3 | NP_062512.1 | 2 | 423 | ||
GenBank mRNA | AB025943.1 | - | 1 | 281 | ||
AK143623.1 | - | 2 | 416 | |||
AK149848.1 | - | 2 | 416 | |||
AK169853.1 | - | 2 | 407 | |||
BC059775.1 | - | 2 | 401 | 5370916 | ||
BQ942269.1 | - | 2 | 423 | |||
EF524070.1 | - | 2 | 375 |