Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019571.5 | NP_062517.1 | 3 | 588 | ||
GenBank mRNA | AF121344.1 | - | 3 | 603 | ||
AK013350.1 | - | 3 | 553 | |||
AK015705.1 | - | 3 | 310 | |||
AK033543.1 | - | 3 | 287 | |||
AK187703.1 | - | 2 | 102 | |||
AK210177.1 | - | 2 | 122 | |||
BC046784.1 | - | 3 | 491 | 6416107 | ||
BC058695.1 | - | 3 | 559 | 6813872 |